Product Image
Thermo Scientific™
Oxichrome Reagent
Rapidly detect the enzyme cytochrome oxidase in microorganisms using Oxichrome Reagent.
Have Questions?
Catalog NumberQuantity
R830900915 mL/Vial
Catalog number R8309009
Price (USD)
-
Quantity:
15 mL/Vial
Rapidly detect the enzyme cytochrome oxidase in microorganisms using Thermo Scientific™ Remel™ Oxichrome Reagent. The oxidase test was originally developed to identify Neisseria spp., but later was used to separate pseudomonads from oxidase-negative enteric Gram-negative bacilli (Enterobacteriaceae)1,2. Most Gram-positive bacteria are oxidase-negative, pseudomonads are oxidase-positive, and other non-enteric, Gram-negative bacilli, are variable
Pseudomonads, bacteria of the family Pseudomonadaceae are a large and varied group comprising four major genera and several hundred species. Most species of the family are found in soil or water; some cause diseases in plants, and a few cause serious diseases in humans and other mammals. Pseudomonas aeruginosa, very common and widespread, is an opportunistic pathogen for man that causes antibiotic-resistive infections in persons of weakened resistance.
The oxidase reaction is attributed to the presence of a cytochrome oxidase system in the respiratory chain of certain bacteria, which utilizes molecular oxygen as a terminal electron acceptor3,4. Oxidase-positive bacteria produce oxidase enzymes which, in the presence of atmospheric oxygen, cytochrome c, and an oxidase reagent (Oxichrome Reagent), oxidize the reagent to form a blue-purple colored compound, Wurster’s blue.
  • Ready-to-use – Liquid reagent
  • Easy to interpret – Blue to purple color indicates a positive test
  • Easy-to-store – Storage at room temperature
  • Rapid results – Can be interpreted within 30 seconds
Oxichrome reagent contains N,N,N,N-tetramethyl-ρ-phenylenediamine supplied in 15 mL bottles.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
  1. Kovacs, N. 1956. Nature. 178:703.
  2. Cowan, S.T. and K.J. Steel. 1965. Manual for the Identification of Medical Bacteria. Cambridge University Press, Cambridge, UK.
  3. Steel, K.J. 1961. J. Gen. Microbiol. 25:297-306.
  4. MacFaddin, J.F. 2000. Biochemical Tests for Identification of Medical Bacteria. 3rd ed. Lippincott Williams & Wilkins, Philadelphia, PA.
Specifications
DescriptionOxichrome Reagent
FormatVial
Packaging Type15 mL/Vial
Quantity15 mL/Vial
Unit SizeEach
Contents & Storage
Store at Room Temperature Until Used