Can I buy your anti-epitope antibodies in larger aliquots than 50 ul?
Yes, some of our unconjugated antibodies can be purchased in 500 µl aliquots. The catalog numbers are: R910-CUS (Anti-Xpress antibody), R930-CUS (Anti-His C-term antibody), R940-CUS (Anti-HisG antibody), R950-CUS (Anti-myc antibody), R960-CUS (Anti-V5 antibody). These are considered custom sizes and are not always available for immediate shipment, so please be aware that there may be an extra lead time for your order.