Anti-Thio™ Antibody - FAQs
Anti-Thio™ Antibody - FAQs
View additional product information for Anti-Thio™ Antibody - FAQs (R92025)
1 product FAQs found
Does the Anti-Thio antibody detect the His-Patch in the His-Patch ThioFusion System?
Yes, the His-Patch vectors have been tested successfully with the Anti-Thio antibody in Western blot analysis.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.