Anti-His(C-Term) Antibody - FAQs
Anti-His(C-Term) Antibody - FAQs
View additional product information for Anti-His(C-Term) Antibody - FAQs (R930CUS)
1 product FAQs found
Can I buy your anti-epitope antibodies in larger aliquots than 50 ul?
Yes, some of our unconjugated antibodies can be purchased in 500 µl aliquots. The catalog numbers are: R910-CUS (Anti-Xpress antibody), R930-CUS (Anti-His C-term antibody), R940-CUS (Anti-HisG antibody), R950-CUS (Anti-myc antibody), R960-CUS (Anti-V5 antibody). These are considered custom sizes and are not always available for immediate shipment, so please be aware that there may be an extra lead time for your order.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.