HisG Epitope Tag Monoclonal Antibody, AP - FAQs
HisG Epitope Tag Monoclonal Antibody, AP - FAQs
View additional product information for HisG Epitope Tag Monoclonal Antibody, AP - FAQs (R94225)
2 product FAQs found
Can the Anti-His (C-term) antibody be used against the N-terminal HisG (His-His-His-His-His-His-Gly) epitope tag?
You cannot use Anti-His (C-term) antibody against the N-terminal HisG epitope taq. Thermo Fisher Scientific sells two Anti-His antibodies: Anti-HisG and Anti-His (C-term) antibodies depending on the epitope tag in your vector. These antibodies do not recognize react well with both epitopes because the glycine and the carboxy terminus are important parts of each one. Any 6xHis tag lacking in these additional elements is a poor epitope for either of the antibodies.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What applications are appropriate for your Anti-Xpress antibody, Anti-V5 antibody, Anti-HisG antibody, and Anti-myc antibody?
ELISA, Western blot, Immunoprecipitation, Immunofluorescence, and Immunohistochemistry protocols have all been used successfully with our epitope tag antibodies. Please consult the product manuals for specific protocols and dilutions.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.