GFAP (Glial Fibrillary Acidic Protein) Rabbit Polyclonal Antibody, Epredia™ - FAQs
GFAP (Glial Fibrillary Acidic Protein) Rabbit Polyclonal Antibody, Epredia™ - FAQs
View additional product information for GFAP (Glial Fibrillary Acidic Protein) Rabbit Polyclonal Antibody, Epredia™ - FAQs (RB-087-A1)
1 product FAQs found
Do you offer primary antibodies that are specific for neurons or glial cells?
Please note that only antibodies against neuronal proteins are specific to neuronal cells. We carry antibodies to GFAP, HuC/HuD, NMDA receptor, synapsin I as well as many other antibodies for neuronal proteins. A link to our primary antibody search tool can be found:
Neurobiology antibody search (https://www.thermofisher.com/us/en/home/life-science/antibodies/primary-antibodies/neurobiology-antibodies.html)