Human Insulin Recombinant Protein - FAQs
Human Insulin Recombinant Protein - FAQs
View additional product information for Human Insulin Recombinant Protein - FAQs (RP10935, RP10908)
1 product FAQs found
What are insulin analogs?
There are 4 kinds of insulin analogs:
  • NPH insulin (Humulin N)
  • Lispro insulin (Humalog)
  • Insulin Aspart (Novolog)
  • Insulin glulisine