Liver S9 fractions prepared from a pool of male Sprague Dawley rats. Total protein concentration: 20mg/mL
Have Questions?
Catalog Number
Quantity
RTS9PL
1 mL
Catalog number RTS9PL
Price (USD)
55.65
Online exclusive
75.75
Save 20.10
Each
Quantity:
1 mL
Price (USD)
55.65
Online exclusive
75.75
Save 20.10
Each
Liver S9 fractions prepared from a pool of male Sprague Dawley rats.
Total protein concentration: 20 mg/ml
We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.
For research use only. Not intended for human or animal therapeutic or diagnostic use.