Human Keratinocyte Growth Supplement (HKGS) - FAQs
Human Keratinocyte Growth Supplement (HKGS) - FAQs
View additional product information for Human Keratinocyte Growth Supplement (HKGS) - FAQs (S0015)
1 product FAQs found
Do I need to add additional supplements to AmnioMAX-II Complete Medium (Cat. No. 11269016)?
No. AmnioMax-II Complete Medium (Cat. No. 1129016) is fully supplemented with antibiotics, L-glutamine, and FBS.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.