Attachment Factor Protein (1X) - FAQs
Attachment Factor Protein (1X) - FAQs
View additional product information for Attachment Factor Protein (1X) - FAQs (S006100)
1 product FAQs found
What is the source of the gelatin used in Attachment Factor Protein (1X)?
The gelatin is derived from porcine skin.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.