How many binding sites does the streptavidin within the Streptavidin, Alexa Fluor 568 conjugate (Cat. No S11226) have and what is its molecular weight?
Each streptavidin molecule within the Streptavidin, Alexa Fluor 568 conjugate (Cat. No S11226) has 2-4 binding sites, which means that it can bind 2-4 dye molecules. However, the exact number of binding sites is lot-dependent and can be found on the lot-specific Certificate of Analysis.
The molecular weight (MW) of streptavidin is 52.8 kDa and each Alexa Fluor 568 dye molecule has a MW of 679 Da, so the MW of the Streptavidin, Alexa Fluor 568 conjugate is in the range of 54.2-55.5 kDa.