SYPRO™ Ruby Protein Blot Stain - FAQs
SYPRO™ Ruby Protein Blot Stain - FAQs
View additional product information for SYPRO™ Ruby Protein Blot Stain - FAQs (S11791)
1 product FAQs found
I have stained my blot with Pro-Q Diamond Phosphoprotein Blot Stain. Can I also stain the blot with SYPRO Ruby Blot Stain for total protein detection?
Yes. We recommend staining with SYPRO Ruby Blot Stain after staining with Pro-Q Diamond Phosphoprotein Blot Stain.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.