DNA Sequencing Systems: Combs for Discontinued Model FB-SEQ-2045
Actual product may vary
DNA Sequencing Systems: Combs for Discontinued Model FB-SEQ-2045
DNA Sequencing Systems: Combs for Discontinued Model FB-SEQ-2045
DNA Sequencing Systems: Combs for Discontinued Model FB-SEQ-2045
DNA Sequencing Systems Combs for discontinued model FB-SEQ-2045
Have Questions?
Catalog NumberThickness (Metric) TeethVolume (Metric) WellProduct TypeTeeth Count
S1S-20A0.4 mm15μLSharktooth Comb20
Catalog number S1S-20A
Price (USD)
59.40
Online exclusive
66.00
Save 6.60
Each
Thickness (Metric) Teeth:
0.4 mm
Volume (Metric) Well:
15μL
Product Type:
Sharktooth Comb
Teeth Count:
20
Price (USD)
59.40
Online exclusive
66.00
Save 6.60
Each
For use with discontinued model FB-SEQ-2045
Specifications
DescriptionWell Comb
For Use With (Equipment)Owl s4s aluminum backed sequencer system
Height (English) Footprint19.6 in.
Height (Metric) Footprint50 cm
Length (English) Footprint9 in.
Length (English) Gel17.7 in.
Length (Metric) Footprint23 cm
Length (Metric) Gel45 cm
Product TypeSharktooth Comb
Teeth Count20
Thickness (Metric) Teeth0.4 mm
Volume (Metric) Well15μL
Width (English) Footprint10.6 in.
Width (Metric) Footprint27 cm
Width (Metric) Gel20 cm
Unit SizeEach