QuEChERS EN 15662 Method Clean-up Kits
Thermo Scientific™

QuEChERS EN 15662 Method Clean-up Kits

Use Thermo Scientific™ QuEChERS EN 15662 Method Clean-up Kits to analyze complex sample matrices for residues, toxins, and other contaminants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample TypeTube Size
S2-2-HP-EN-KITHighly Pigmented Fruits and Vegetables2 mL
S2-15-FW-EN-KITFruits and Vegetables with Fats and Waxes15 mL
S2-15-HP-EN-KITHighly Pigmented Fruits and Vegetables15 mL
S2-15-P-EN-KITPigmented Fruits and Vegetables15 mL
S2-2-FW-EN-KITFruits and Vegetables with Fats and Waxes2 mL
S2-2-GFV-EN-KITGeneral Fruits and Vegetables2 mL
S2-2-P-EN-KITPigmented Fruits and Vegetables2 mL
Catalog number S2-2-HP-EN-KIT
Price (USD)
283.55
Pack of 100
Sample Type:
Highly Pigmented Fruits and Vegetables
Tube Size:
2 mL
Price (USD)
283.55
Pack of 100
Use Thermo Scientific™ QuEChERS Clean-up Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the EN 15662 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

Thermo Scientific™ QuEChERS EN 15662 Method Clean-Up Kits make sample preparation straight-forward and efficient, enabling you to confidently perform extractions and analyze fruit and vegetable samples for pesticide residues and other contaminants.

Clean-up kits are available in prefilled 2 mL and 15 mL tube formats for the following matrices: general fruits and vegetables, pigmented fruits and vegetables, highly pigmented fruits and vegetables, and fruits and vegetables containing fats and waxes.

What is the purpose of the chemicals used in the EN 15662 Method?

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer; removes remaining water from sample
  • NaCl: induces phase separation between water content in sample and acetonitrile layer; removes residual water from sample
  • Trisodium Citrate: buffers the sample to stabilize pH
  • Disodium Citrate Sesquihydrate: buffers the sample to stabilize pH
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids, from sample
  • C18: removes fats, sterols, and other non-polar interferences from sample
  • GCB: (graphitized carbon black) removes pigment (not recommended for use with planar pesticides)
Specifications
DescriptionQuEChERS EN 15662 Method Clean-up Kit; 150 mg MgSO4, 25 mg PSA, 7.5 mg GCB prefilled in 2 mL Tubes; 100/Pk.
IncludesSalts in Prefilled Tubes
MethodQuEChERS sample preparation for EN 15662 Method
Salt Composition150 mg MgSO4, 25 mg PSA, 7.5 mg GCB
Sample TypeHighly Pigmented Fruits and Vegetables
Tube Size2 mL
TypeQuEChERS Clean-up Kit
Unit SizePack of 100