SAIVI™ Alexa Fluor™ 647 Antibody/Protein 1 mg-Labeling Kit - FAQs
SAIVI™ Alexa Fluor™ 647 Antibody/Protein 1 mg-Labeling Kit - FAQs
View additional product information for SAIVI™ Alexa Fluor™ 647 Antibody/Protein 1 mg-Labeling Kit - FAQs (S30044)
4 product FAQs found
What amount of conjugated antibody should I inject to image tumors?
A recommended starting dosage is 50 µg. You will need to determine the optimal dosage for your experimental model.
What size needle should I use for small animal in vivo imaging?
We recommend the use of a 28-32 gauge tuberculin or insulin syringe (0.3 or 1.0 mL volume) with a fixed (non-removable) needle.
What are the optimal and maximal volumes of reagent that can be injected into a mouse?
The volume of reagent that can be injected varies according to the route of administration. The following numbers are general guidelines for a 25 gram animal: Intravenous (IV)- 50-125 µl (recommended)- 200µl (maximum) ; Intraperitoneal (IP) 500µl (recommended) -2ml (maximum) ; Subcutaneous(SC) 100-250 µl (recommended)- 1ml (maximum).
What type of mice are the best for in vivo imaging?
Due to light scattering caused by fur, hairless mice such as athymic nude (nu/nu) mice are recommended for in vivo imaging. If this is not an option, the hair covering the area to be imaged should be removed using clippers or a chemical depilatory such as Nair depilatory.