SYTO™ RNASelect™ Red
Invitrogen™

SYTO™ RNASelect™ Red

SYTO™ RNASelect™ Red is a cell-permeant, RNA-selective fluorescent dye for live or fixed eukaryotic cells. It allows researchers to visualize RNA-rich nucleoli and cytoplasmic RNA in a Texas Red/RFP compatible channel.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
S327113 x 300 μL
S32710300 μL
Catalog number S32711
Price (USD)
495.00
Each
Quantity:
3 x 300 μL
Price (USD)
495.00
Each

SYTO™ RNASelect™ Red is a cell-permeant, RNA-selective fluorescent dye for live or fixed eukaryotic cells. It allows researchers to visualize RNA-rich nucleoli and cytoplasmic RNA in a Texas Red/RFP compatible channel. Compared to SYTO™ RNASelect™ Green, it is compatible with 4% paraformaldehyde fixation rather than methanol fixation, and leaves GFP/FITC channels open for antibodies, fluorescent proteins, and other fluorescent probes in multiplex and high-content imaging.

This molecular probe is prepared as a ready-to-use 100x aqueous solution, appropriate for direct addition to either complete media or other buffers of your choice to then be used in staining cells.

Compatibility with 4% paraformaldehyde fixation and permeabilization expands the applicability of this RNA dye for immunofluorescence workflows, enabling its use in high content screening for nucleolar reorganization, cell stress assays, and deep learning computational models.

The staining patterns demonstrate high RNA specificity, with expected staining demonstrating bright nucleoli and diffuse ribosomal staining throughout the cytoplasm and is easily multiplexable with classic nuclear stains such as Hoechst 33342 or DAPI. This specificity is demonstrated by the sharp decrease in staining intensity upon treatment with RNase, all red signal is quickly depleted while the nuclear DNA dye staining remains intact.

Features

  • Suitable for both live and fixed cell workflows, connecting live cell observation with endpoint imaging.
  • Compatible with multiplex imaging outside of the Texas Red/RFP channels with various organelle markers such as Hoechst/DAPI, phalloidin, ER, lectins, or mitochondrial probes.
  • Applicable to high-content imaging and RNA based nucleolar morphology studies, including workflows where nucleolar signal is used for segmentation or comparative phenotyping.

For researchers familiar with SYTO™ 14, SYTO™ RNASelect™ Red offers a red channel option for RNA associated morphology in Cell Painting related or other multiplex imaging workflows.

This dye complements SYTO™ RNASelect™ Green in channel planning and fixation choice: SYTO™ RNASelect™ Green remains the better fit when the green channel is desired and the sample type requires methanol fixation, whereas SYTO™ RNASelect™ Red is appropriate when a red RNA channel or 4% PFA workflow is preferred.

Available as a single vial as S32710 and in a 3-pack format as S32711.

For Research Use Only.
Specifications
Cell PermeabilityLive and Fixed Cell Permeable
ColorRed
Concentration1 mM in 25% DMSO/DI H2O
Detection MethodFluorescence with Texas Red Filter Set
Dye TypeFluorogenic Organic Dye
Emission626 nm
Excitation Wavelength Range595 nm
For Use With (Application)Live Cell Imaging, Fixed Cell Imaging, High Content Screening, Fluorescence Microscopy
For Use With (Equipment)Fluorescence Microscope, Spectral Imaging Instrument, Flow Cytometer, High Content Imager
FormAqueous Liquid with 25% DMSO
FormatVial
No. of Tests900 Tests
Product LineSYTO RNASelect
Quantity3 x 300 μL
ReactivityRNA, Nucleolar Structure
Reagent TypeFluorogenic RNA Label
TargetRNA
TechniqueHigh Content Imaging, Fluorescence Microscopy
Validated ApplicationLive Cell Imaging, Fixed Cell Imaging, High Content Imaging
Volume (Metric)900 μL
Label TypeFluorescent Dye
Product TypeRNA-specific Label
Sub Cellular LocalizationNucleolus
Unit SizeEach
Contents & Storage
•3 Vials
•Store at 2°C to 30°C
•Protect from light
•Do not freeze

Frequently asked questions (FAQs)

The process of purifying exosomes using ultra-centrifugation differs greatly among published papers. How can I determine which protocol is most suitable for my cell line?

There are some variations to the ultracentrifugation protocols, not based on the cell lines used so much as the experience in that particular lab, including what G-force they recommend, duration of ultra-centrifugation, straight sedimentation vs cushion vs sucrose gradients to obtain the top quality exosomes at reasonable yields. We often recommend protocols developed by Clothilde: C. Thery, S. Amigorena, G. Raposo and A. Clayton “Isolation and characterization of exosomes from cell culture supernatants and biological fluids.” Curr. Protoc. Cell. Biol., Chapter 3, Unit 3: 22, 2006.

Can I use PBS with SYTO RNASelect Red, SYTO RNASelect Green, and SYTO 14 reagents?

PBS can be acceptable in some workflows, but we do not recommend assuming it is always the best-performing buffer for all three probes. Some applications may perform better in phosphate-free buffers, so we suggest optimizing as needed. When a starting point is needed, we recommend Live Cell Imaging Solution (LCIS; Cat. No. A59688DJ).

Can I use SYTO RNASelect Red, SYTO RNASelect Green, or SYTO 14 reagents as absolute measurements of total RNA content?

We recommend using these stains as relative imaging reagents within a controlled workflow rather than as absolute measurements of total RNA content. SYTO RNASelect Green reagent has been used in quantitative RNA content assay workflows, but any quantitative application should be carefully calibrated and ideally confirmed with an orthogonal RNA quantitation method.

Are SYTO RNASelect Red, SYTO RNASelect Green, and SYTO 14 reagents transcript-specific?

No. We recommend thinking of SYTO RNASelect Red, SYTO RNASelect Green, and SYTO 14 reagent as RNA-associated stains rather than sequence-targeted probes.

Can I use SYTO RNASelect Red, SYTO RNASelect Green, or SYTO 14 reagents for automated segmentation or machine-learning analysis?

Yes. We suggest these dyes can be used for automated segmentation and machine-learning analysis, depending on the workflow. SYTO 14 reagent has strong Cell Painting precedent, and SYTO RNASelect Red reagent has shown utility in high-content imaging workflows that rely on segmentation-oriented nucleolar analysis.