What is the size range of exosomes that can be isolated using the Total Exosome Isolation Kit (from serum)?
By Nanosight analysis, most of the recovered exosomes are in the expected size range, 30-150 nm.
Search
View additional product information for SYBR GREEN I AND SYBR GREEN II - FAQs (S32717)
1 product FAQs found