PRODUCT INFO SHEETS FOR S7580 - FAQs
PRODUCT INFO SHEETS FOR S7580 - FAQs
View additional product information for PRODUCT INFO SHEETS FOR S7580 - FAQs (S32718)
1 product FAQs found
What is the best way to resuspend the pellet of exosomes when using your Total Exosome Isolation kit?
The pellet is soluble in PBS or any other buffer. It is very easy to resuspend exosomes recovered from a small sample of cell culture medium (e.g., 1 mL), but if you are processing a large sample of plasma or serum, the pellet will be pretty large. In this case, you can add buffer, let it sit for 30 min at 37 degrees C, then vortex gently or pipet up and down to resuspend the exosomes.