SYBR Safe DNA gel stain was developed specifically for reduced mutagenicity – to be safer than ethidium bromide for stainingRead more
Have Questions?
Change view
Catalog Number
Quantity
S33100
1 L
S33101
4 L
2 Options
Catalog number S33100
Price (USD)
157.65
Online exclusive
171.00
Save 13.35
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
157.65
Online exclusive
171.00
Save 13.35
Each
SYBR Safe DNA gel stain was developed specifically for reduced mutagenicity – to be safer than ethidium bromide for staining DNA in agarose or acrylamide gels. SYBR Safe stain is not only less mutagenic than ethidium bromide, but SYBR Safe stain's detection sensitivity is better than that of ethidium bromide. SYBR Safe stain comes as a premixed solution that can be used just like an ethidium bromide solution, either in the gel during the electrophoresis run or as a post-run stain. Bound to nucleic acids, the green-fluorescent SYBR Safe stain has fluorescence excitation maxima at ∼280 and ∼502 nm, and an emission maximum at ∼530 nm. SYBR Safe DNA gel stain is also available as 1 L (S-33100) of ready-to-use solution. The SYBR Safe DNA Gel Stain Starter Kit (S-33110) includes 1 L of SYBR Safe DNA gel stain and one photographic filter (S-37100).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection LocationIn-Gel Detection
Detection MethodFluorescence
Quantity1 L
Shelf Life6 months from date of purchase
Shipping ConditionRoom Temperature
Target MoleculeDNA
Label or DyeSYBR Safe
Product TypeDNA Gel Stain
Unit SizeEach
Contents & Storage
• Supplied as a ready-to-use solution in 0.5X TBE
Store at room temperature in the original container.
Frequently asked questions (FAQs)
Why do I sometimes see speckles in my gel when using SYBR Safe DNA Gel Stain?
Many whitening agents used in clothing, as well as some fungi and bacteria, fluoresce at the same wavelengths as SYBR Safe DNA gel stain. These contaminants within or on the surface of the gel may produce this speckling.
What is the pH range of SYBR dyes?
The SYBR dyes are useful only over a narrow range of pH, from about 7 to 8. Outside this range, the fluorescent signal diminishes rapidly.
Which direction does the SYBR Safe dye run during electrophoresis?
Similarly to ethidium bromide, SYBR Safe DNA Gel Stain runs in the opposite direction of the migrating DNA. This has no practical effect on the use of gels cast with SYBR Safe DNA Gel Stain, as only the very bottom of the gel will have a lower concentration of stain. This effect can be partially counteracted by staining the gel with SYBR Safe DNA Gel Stain after electrophoresis. Solutions of dye should not be added to the running buffer as this can cause breakdown of the dye at the electrodes and release toxic volatile compounds into the air.
Does ethanol precipitation remove the SYBR Safe dye?
SYBR Safe DNA Gel Stain is easily removed from nucleic acids by ethanol precipitation.
Can I reuse SYBR Safe DNA Gel Stain for a second gel?
We strongly discourage the reuse of SYBR Safe DNA Gel Stain, as this practice significantly lowers sensitivity.
Features of medullary thymic epithelium implicate postnatal development in maintaining epithelial heterogeneity and tissue-restricted antigen expression.
Authors:Gillard GO, Farr AG
Journal:J Immunol
PubMed ID:16670287
'Although putative thymic epithelial progenitor cells have been identified, the developmental potential of these cells, the extent of medullary thymic epithelium (mTEC) heterogeneity, and the mechanisms that mediate the expression of a wide range of peripheral tissue-restricted Ags (TRAs) by mTECs remain poorly defined. Here we have defined several basic ... More
Biotinylated photocleavable polyethylenimine: capture and triggered release of nucleic acids from solid supports.
Authors:Handwerger RG, Diamond SL
Journal:Bioconjug Chem
PubMed ID:17432825
'A biotinylated photocleavable polyethylenimine (B-PC-PEI) was designed and synthesized for the capture and controlled release of nucleic acids from solid supports. B-PC-PEI was synthesized via a three-step reaction process and verified by 1H NMR and mass spectrometry. In aqueous solution, the o-nitrobenzyl group within B-PC-PEI was efficiently cleaved by 5 ... More
Development of a SYBR safe technique for the sensitive detection of DNA in cesium chloride density gradients for stable isotope probing assays.
Authors:Martineau C, Whyte LG, Greer CW,
Journal:J Microbiol Methods
PubMed ID:18329741
SYBR safe, a fluorescent nucleic acid stain, was evaluated as a replacement for ethidium bromide (EtBr) in cesium chloride (CsCl) density gradients for DNA stable isotope probing (DNA-SIP) assays. The separation of 12C- and 13C-labelled DNA using SYBR safe gave similar results to those obtained using EtBr with pure cultures ... More
Single step protocol to purify recombinant proteins with low endotoxin contents.
Authors:Reichelt P, Schwarz C, Donzeau M
Journal:Protein Expr Purif
PubMed ID:16290005
Endotoxin is an unwanted by product of recombinant proteins purified from Escherichia coli. The inherent toxicity of endotoxins makes their removal an important step for the proteins' application in several biological assays and for safe parenteral administration. The method described in this paper is a one-step protocol which is effective ... More
5-Methyltetrahydrofolate inhibits photosensitization reactions and strand breaks in DNA.
Authors:Offer T, Ames BN, Bailey SW, Sabens EA, Nozawa M, Ayling JE
Journal:FASEB J
PubMed ID:17341682
The known functions of folate are to support one-carbon metabolism and to serve as photoreceptors for cryptochromes and photolyases. We demonstrate that 5-methyltetrahydrofolate (5-MTHF, the predominant folate in plasma) is also a potent, near diffusion limited, scavenger of singlet oxygen and quencher of excited photosensitizers. Both pathways result in decomposition ... More