SYTO™ Green Fluorescent Nucleic Acid Stains
Actual product may vary
Invitrogen™

SYTO™ Green Fluorescent Nucleic Acid Stains

SYTO Green Fluorescent Nucleic Acid Stains exhibit bright, green fluorescence upon binding to nucleic acids. These dyes are cell-permeant and can be used to stain RNA and DNA in live cells, dead cells, and bacteria.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDye TypeQuantity
S34855SYTO BC Green100 μL
S34854SYTO 9 Green100 μL
S7573SYTO 11 Green250 μL
S7574SYTO 12 Green315 μL
S7575SYTO 13 Green250 μL
S7576SYTO 14 Green250 μL
S7578SYTO 16 Green250 μL
S7556SYTO 21 Green250 μL
S7559SYTO 24 Green250 μL
S32703SYTO RNASelect Green100 μL
S7572SYTO Green Dyes 11-14, 16, 21, 24, and 251 Kit (50 μL each)
Catalog number S34855
Price (USD)
296.65
Online exclusive
395.00
Save 98.35
Each
Dye Type:
SYTO BC Green
Quantity:
100 μL
Price (USD)
296.65
Online exclusive
395.00
Save 98.35
Each

The SYTO Green Fluorescent Nucleic Acid Stains exhibit bright, green fluorescence upon binding to nucleic acids. These dyes are cell-permeant and can be used to stain RNA and DNA in live cells, dead cells, and bacteria. Because the dye characteristics and staining pattern of the SYTO dyes may vary, we offer 10 different SYTO Green Fluorescent Nucleic Acid Stains available individually along with the SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit #1 (containing SYTO 11–14, 16, 21, 24, and 25) to enable researchers to find the most appropriate green-fluorescent SYTO stain for their system . We also provide the SYTOX Green Nucleic Acid Stain (Cat. No. S7020 and R37109), which is a cell-impermeant green-fluorescent stain used for staining dead cells or performing nuclear counterstaining in fixed cells.

SYTO Green Fluorescent Nucleic Acid Stains are cell-permeant nucleic acid stains that show a large fluorescence enhancement upon binding nucleic acids. The SYTO dyes can be used to stain RNA and DNA in both live and dead eukaryotic cells, as well as in Gram-positive and Gram-negative bacteria.

The SYTO stains share several important characteristics:

  • Permeability to virtually all cell membranes, including mammalian cells and bacteria
  • High molar absorptivity, with extinction coefficients >50,000 cm-1 M-1 at visible absorption maxima
  • Extremely low intrinsic fluorescence, with quantum yields typically <0.01 when not bound to nucleic acids
  • Quantum yields that are typically >0.4 when bound to nucleic acids.

SYTO Green-Fluorescent Nucleic Acid Stains excitation/emission wavelengths

  • SYTO 9—RNA 486/501 nm; DNA 485/498 nm
  • SYTO 11—RNA 510/530 nm; DNA 508/527 nm
  • SYTO 12—RNA 500/519 nm; DNA 499/522 nm
  • SYTO 13—RNA 491/514 nm; DNA 488/509 nm
  • SYTO 14—RNA 521/547 nm; DNA 517/549 nm
  • SYTO 16—RNA 494/525 nm; DNA 488/518 nm
  • SYTO 21—DNA 494/517 nm
  • SYTO 24—DNA 490/515 nm
  • SYTO BC—RNA 487/504 nm; DNA 485/500 nm
  • SYTO RNASelect—RNA 490/530 nm

SYTO dyes differ from each other in one or more characteristics, including cell permeability, fluorescence enhancement upon binding nucleic acids, excitation and emission spectra, DNA/RNA selectivity, and binding affinity. The SYTO dyes are compatible with a variety of fluorescence-based instruments that use laser excitation or a conventional broadband illumination source (e.g., mercury- and xenon-arc lamps). Filter sets that are suitable for imaging cells labeled with fluorescein (FITC), Alexa Fluor 488, or GFP will work well for imaging cells stained with SYTO Green Nucleic Acid Stains.

SYTO nucleic acid stains have been used in diverse applications from staining DNA spotted on microarrays to staining live and fixed cells. The SYTO dyes do not act exclusively as nuclear stains in live cells and should not be equated with DNA-selective compounds such as DAPI (Cat. Nos. D1306, D21490) or Hoechst 33342 (Cat. Nos. H1399, H3570), which stain nuclei in live animal cells. Eukaryotic cells incubated with SYTO dyes generally show cytoplasmic or mitochondrial staining, as well as nuclear staining. The SYTO Green Fluorescent Nucleic Acid Stains have proven valuable in a broad range of research applications as well as used in flow cytometry and fluorescence microscopy.

Specific staining applications for some of the SYTO Green Fluorescent Nucleic Acid Stains

The SYTO 9 stain has been shown to stain live and dead Gram-positive and Gram-negative bacteria. It is a component of the LIVE/DEAD BacLight Bacterial Viability Kits (Cat. Nos. L7007, L7012, L13152).

The SYTO 11 stain (Cat. No. S7573) has been used in conjunction with time-lapse microscopy to examine the cleavage orientation of dividing cells in the developing cerebral cortex.

The SYTO 13 stain (Cat. No. S7575) has been used in combination with propidium iodide (Cat. Nos. P1304, P3566) to monitor glutamate-induced necrosis in cerebellar granule cells.

The SYTO 14 stain (Cat. No. S7576) binds to cytoplasmic RNA, allowing its use in tracking RNA granule transport in living neurons.

Several reports describe the use of SYTO dyes for detecting apoptosis. A series of SYTO nucleic acid stains was screened for the ability to discriminate between apoptotic and non-apoptotic mouse thymocytes, and the SYTO 16 stain (Cat. no. S7578) was found to be optimal for this application. The SYTO 16 stain has also been used with propidium iodide to differentiate live and dead COS-7 cells with a laser-based scanning cytometer.

The SYTO BC Green Fluorescent Nucleic Acid Stain is a mixture of some of our best SYTO dyes for bacterial staining and is a component of the Bacteria Counting Kit (Cat. No. B7277) for bacterial counting in flow cytometry.

The SYTO RNASelect Green Fluorescent Cell Stain is a cell-permeant nucleic acid stain that selectively stains RNA. Although virtually nonfluorescent in the absence of nucleic acids, the SYTO RNASelect stain exhibits bright green fluorescence when bound to RNA (absorption/emission maxima ∼490/530 nm), but only a weak fluorescent signal when bound to DNA. It has been used for staining RNA cargo in exosomes in both live cell samples and isolated exosomes.

SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit

The SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit #1 contains our collection of cell-permeant, green-fluorescent SYTO nucleic acid stains. Because the dyes may demonstrate different staining behaviors with various tissues and cells, it may be necessary to test the dyes to find the optimal dye for a specific application. The kit contains 50 μL each of SYTO 11–14, 16, 21, 24, and 25 dyes.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Concentration5 mM
Detection MethodFluorescence
Dye TypeSYTO BC Green
EmissionRNA: 504 nm, DNA: 500 nm
Excitation Wavelength RangeRNA: 487 nm, DNA: 485 nm
For Use With (Application)Fluorescent Labeling, Live Cell Imaging, Microarray
For Use With (Equipment)Fluorescence Microscope, High Content Imager, Flow Cytometer
FormSolution
Product LineSYTO
Quantity100 μL
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
Volume (Metric)100 μL
Label TypeFluorescent Dye
Product TypeNucleic Acid Stain
Sub Cellular LocalizationNucleic Acids
Unit SizeEach
Contents & Storage
Contains 1 vial SYTO™ BC (100 μL, 5 mM in DMSO).

Store in freezer (-5°C to -30°C) and protect from light.

Frequently asked questions (FAQs)

Is the SYTO BC bacteria stain from the Bacteria Counting Kit available as a standalone product?

Yes, the SYTO BC bacteria stain can be purchased separately (Cat. No. S34855).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How do SYTO dyes bind to DNA?

The binding mode of SYTO nucleic acid stains is unknown. However, the behavior of these and related nucleic acid dyes suggests the following binding properties:

1.They appear to contact the solvent (suggested by sensitivity to salt, divalent cations, and in particular, SDS) and thus are likely to have contacts in the grooves.
2.All SYTO dyes appear to show some base selectivity and are thus likely to have minor groove contacts.
3.They can be removed from nucleic acid via ethanol precipitation; this characteristic is not shared by ethidium bromide and other intercalators. Likewise, the dyes are not removed from nucleic acid via butanol or chloroform extraction. These extraction methods do remove ethidium bromide from nucleic acid. 4. SYTO binding is not affected by nonionic detergents.
5. SYTO dyes are not quenched by BrdU, so they do not bind nucleic acids in precisely the same way as Hoechst 33342 and DAPI ((4′,6-diamidino-2-phenylindole).

SYBR Green I has shown little mutagenicity on frameshift indicator strains, indicating that it isn't likely to strongly intercalate.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Do you have spectra information for SYTO BC Green Fluorescent Nucleic Acid Stain (Cat. No. S34855)?

Spectra information for SYTO BC Green Fluorescent Nucleic Acid Stain can be found on page 5 in the user manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/mp07572.pdf).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (12)

Citations & References
Abstract
A flow-cytometric gram-staining technique for milk-associated bacteria.
Authors:Holm C, Jespersen L
Journal:Appl Environ Microbiol
PubMed ID:12732558
'A Gram-staining technique combining staining with two fluorescent stains, Oregon Green-conjugated wheat germ agglutinin (WGA) and hexidium iodide (HI) followed by flow-cytometric detection is described. WGA stains gram-positive bacteria while HI binds to the DNA of all bacteria after permeabilization by EDTA and incubation at 50 degrees C for 15 ... More
Endopeptidase penicillin-binding proteins 4 and 7 play auxiliary roles in determining uniform morphology of Escherichia coli.
Authors:Meberg BM, Paulson AL, Priyadarshini R, Young KD,
Journal:J Bacteriol
PubMed ID:15576782
'The low-molecular-weight (LMW) penicillin-binding protein, PBP 5, plays a dominant role in determining the uniform cell shape of Escherichia coli. However, the physiological functions of six other LMW PBPs are unknown, even though the existence and enzymatic activities of four of these were established three decades ago. By applying fluorescence-activated ... More
Genetic diversity of viable, injured, and dead fecal bacteria assessed by fluorescence-activated cell sorting and 16S rRNA gene analysis.
Authors:Ben-Amor K, Heilig H, Smidt H, Vaughan EE, Abee T, de Vos WM,
Journal:Appl Environ Microbiol
PubMed ID:16085863
'A novel approach combining a flow cytometric in situ viability assay with 16S rRNA gene analysis was used to study the relationship between diversity and activity of the fecal microbiota. Simultaneous staining with propidium iodide (PI) and SYTO BC provided clear discrimination between intact cells (49%), injured or damaged cells ... More
Roles of cellular activation and sulfated glycans in Haemophilus somnus adherence to bovine brain microvascular endothelial cells.
Authors:Behling-Kelly E, Vonderheid H, Kim KS, Corbeil LB, Czuprynski CJ,
Journal:Infect Immun
PubMed ID:16926425
'Haemophilus somnus can cause a devastating fibrinopurulent meningitis with thrombotic vasculitis and encephalitis in cattle. The mechanisms used by H. somnus to migrate from the bloodstream into the central nervous system (CNS) are unknown. In this study, we demonstrate that H. somnus adheres to, but does not invade, bovine brain ... More
Space flight alters bacterial gene expression and virulence and reveals a role for global regulator Hfq.
Authors:Wilson JW, Ott CM, Höner zu Bentrup K, Ramamurthy R, Quick L, Porwollik S, Cheng P, McClelland M, Tsaprailis G, Radabaugh T, Hunt A, Fernandez D, Richter E, Shah M, Kilcoyne M, Joshi L, Nelman-Gonzalez M, Hing S, Parra M, Dumars P, Norwood K, Bober R, Devich J, Ruggles A, Goulart C, Rupert M, Stodieck L, Stafford P, Catella L, Schurr MJ, Buchanan K, Morici L, McCracken J, Allen P, Baker-Coleman C, Hammond T, Vogel J, Nelson R, Pierson DL, Stefanyshyn-Piper HM, Nickerson CA,
Journal:Proc Natl Acad Sci U S A
PubMed ID:17901201
'A comprehensive analysis of both the molecular genetic and phenotypic responses of any organism to the space flight environment has never been accomplished because of significant technological and logistical hurdles. Moreover, the effects of space flight on microbial pathogenicity and associated infectious disease risks have not been studied. The bacterial ... More