SlowFade™ Glass Soft-set Antifade Mountant, with DAPI - FAQs
SlowFade™ Glass Soft-set Antifade Mountant, with DAPI - FAQs
View additional product information for SlowFade™ Glass Soft-set Antifade Mountant, with DAPI - FAQs (S36919)
1 product FAQs found
Does Slowfade Antifade Mountant require using a nail polish for sealing?
No. Samples mounted with SlowFade mountants need not be sealed; they are intended for immediate viewing. The coverslip may be anchored (to prevent movement while viewing) by applying molten paraffin to three or four spots around the edge of the coverslip.