SlowFade™ Gold Antifade Mountant with DAPI
Invitrogen™

SlowFade™ Gold Antifade Mountant with DAPI

SlowFade Gold Antifade Mountant is a liquid mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
S369395 x 2 mL
S369422 mL
S3693810 mL
Catalog number S36939
Price (USD)
261.58
Special offer
Online exclusive
Ends: 31-Dec-2026
319.00
Save 57.42
Each
Quantity:
5 x 2 mL
Price (USD)
261.58
Special offer
Online exclusive
Ends: 31-Dec-2026
319.00
Save 57.42
Each

SlowFade Gold Antifade Mountant is a liquid mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides. It contains chemical components designed to protect fluorescent dyes from fading (photobleaching) during fluorescence microscopy experiments. SlowFade Gold Antifade Mountant is glycerol-based, with no mixing or curing required, which makes it ideal for immediate viewing of the sample. It comes ready-to-use—just apply a drop to the sample, add a coverslip, and image. It is available with or without DAPI nuclear stain.

Key attributes:
• Protects dyes from fading during imaging
• Ready-to-use liquid, ideal for immediate sample viewing
• Mounted samples are stable for weeks
• Maintains signal strength—little to no quenching
• Ideal for Alexa Fluor dyes

Select the antifade mounting or optical clearing reagent that matches your fluorescence imaging needs ›

SlowFade Gold Antifade Mountant is not recommended for mounting samples containing fluorescent proteins, such as GFP. For superior antifade protection of fluorescent proteins and fluorescent dyes in cultured cells or thin tissue slices, SlowFade Diamond Antifade Mountant is recommended. For high resolution or to image thicker tissue or 3D cell culture with a focal depth of 0–500 μm, try SlowFade Glass Antifade Mountant.

Select best SlowFade antifade reagent for your experimental needs ›

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeDAPI
Product LineSlowFade
Quantity5 x 2 mL
Shipping ConditionRoom Temperature
For Use With (Equipment)Microscope
FormLiquid
Product TypeMounting Media
Solution TypeAnti-fade Mountant
Unit SizeEach
Contents & Storage
Storage at room temperature is recommended but can also be stored frozen (-5° to -30°C). Protect from light.

Frequently asked questions (FAQs)

What is the difference between ProLong and SlowFade antifade reagents?

Our ProLong antifade reagents dispense as a liquid that will solidify upon the evaporation of water. SlowFade antifade reagents remain liquid. If you are going to image right away and then dispose of your sample, you do not need a mountant that hardens, such as the SlowFade reagents. If you wish to archive your slide for more than a day, you will want a mounting medium that hardens (or “cures”). This hardening will limit the off-rates of various dye-conjugated antibodies and provides a better refractive index. Also, there will be a lower diffusion rate of free radicals, limiting photobleaching.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Some antifade mounting media stay as liquid whereas others harden. What is the benefit of having one that hardens?

If you are going to image right away and then dispose of your sample, you probably want a mountant that does not harden. If you wish to archive your slide for more than a day, you want a mountant that hardens (or "cures"). This hardening will slow or prevent off-rate of your dye or conjugate and often produces a better refractive index. Secondary sealing is usually not necessary. Also there will be lower diffusion of free radicals, thus limiting photobleaching.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I do not have epoxy or VALAP to seal the coverslip. Do you offer an alternative coverslip sealant?

We offer ProLong Coverslip Sealant (Cat. No. P56128) that can be used to seal the edges of the coverslip and is compatible with both curing and non-curing mountant. The sealant is easy to apply and is brushed on after the mountant has cured, for long preservation of slides. The product page can be found here.

When using a hard-curing mountant, such as ProLong Antifade Mountant, make sure the mountant is fully cured before applying ProLong Coverslip Sealant. Since this sealant can seal moisture under the coverslip, it can interfere with the curing process.

When using a non-curing mountant, such as SlowFade Antifade Mountants, sealing can take place immediately after mounting.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Does Slowfade Antifade Mountant require using a nail polish for sealing?

No. Samples mounted with SlowFade mountants need not be sealed; they are intended for immediate viewing. The coverslip may be anchored (to prevent movement while viewing) by applying molten paraffin to three or four spots around the edge of the coverslip.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

DAPI and Hoechst dyes are quite similar to each other. Why would I choose one over the other?

DAPI is a very common blue-fluorescent dye for nuclear counterstaining and gives very bright labeling on nuclei in fixed and permeabilized cells and tissues. However, it is considered to be a semi-permeant to impermeant stain and provides inconsistent staining of live cells. Hoechst 33342 dye is cell-permeant and stains with the same binding mechanism and fluorescent color; it is preferred for live-cell imaging and is just as good as DAPI for fixed cell labeling.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (18)

Citations & References
Abstract
Alkylation damage causes MMR-dependent chromosomal instability in vertebrate embryos.
Authors:Feitsma H, Akay A, Cuppen E,
Journal:Nucleic Acids Res
PubMed ID:18522974
'S(N)1-type alkylating agents, like N-methyl-N-nitrosourea (MNU) and N-ethyl-N-nitrosourea (ENU), are potent mutagens. Exposure to alkylating agents gives rise to O(6)-alkylguanine, a modified base that is recognized by DNA mismatch repair (MMR) proteins but is not repairable, resulting in replication fork stalling and cell death. We used a somatic mutation detection ... More
Identification of a novel signal in the cytoplasmic tail of the Na+:HCO3- cotransporter NBC1 that mediates basolateral targeting.
Authors:Li HC, Li EY, Neumeier L, Conforti L, Soleimani M,
Journal:Am J Physiol Renal Physiol
PubMed ID:17182531
'The Na(+):HCO(3)(-) cotransporter NBC1 (SLC4A4, variant A, kidney specific) is located exclusively on the basolateral membrane of epithelial cells, implying that this molecule has acquired specific signals for targeting to the basolateral membrane. A motif with the sequence QQPFLS (positions 1010-1015) in the cytoplasmic tail of NBC1 was recently demonstrated ... More
Reactive oxygen species production via NADPH oxidase mediates TGF-beta-induced cytoskeletal alterations in endothelial cells.
Authors:Hu T, Ramachandrarao SP, Siva S, Valancius C, Zhu Y, Mahadev K, Toh I, Goldstein BJ, Woolkalis M, Sharma K
Journal:Am J Physiol Renal Physiol
PubMed ID:16159901
'Cytoskeletal alterations in endothelial cells have been linked to nitric oxide generation and cell-cell interactions. Transforming growth factor (TGF)-beta has been described to affect cytoskeletal rearrangement in numerous cell types; however, the underlying pathway is unclear. In the present study, we found that human umbilical vein endothelial cells (HUVEC) have ... More
Identification and characterization of small molecules that inhibit intracellular toxin transport.
Authors:Saenz JB, Doggett TA, Haslam DB
Journal:Infect Immun
PubMed ID:17576758
'Shiga toxin (Stx), cholera toxin (Ctx), and the plant toxin ricin are among several toxins that reach their intracellular destinations via a complex route. Following endocytosis, these toxins travel in a retrograde direction through the endosomal system to the trans-Golgi network, Golgi apparatus, and endoplasmic reticulum (ER). There the toxins ... More
Enhanced innate antiviral gene expression, IFN-a, and cytolytic responses are predictive of mucosal immune recovery during simian immunodeficiency virus infection.
Authors:Verhoeven D, George MD, Hu W, Dang AT, Smit-McBride Z, Reay E, Macal M, Fenton A, Sankaran-Walters S, Dandekar S,
Journal:
PubMed ID:24610016
The mucosa that lines the respiratory and gastrointestinal (GI) tracts is an important portal of entry for pathogens and provides the first line of innate immune defense against infections. Although an abundance of memory CD4(+) T cells at mucosal sites render them highly susceptible to HIV infection, the gut and ... More