SlowFade™ Gold Antifade Mountant
Invitrogen™

SlowFade™ Gold Antifade Mountant

SlowFade Gold Antifade Mountant is a liquid mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
S369402 mL
S369375 x 2 mL
S3693610 mL
Catalog number S36940
Price (USD)
86.65
Online exclusive
93.75
Save 7.10
Each
Quantity:
2 mL
Price (USD)
86.65
Online exclusive
93.75
Save 7.10
Each

SlowFade Gold Antifade Mountant is a liquid mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides. It contains chemical components designed to protect fluorescent dyes from fading (photobleaching) during fluorescence microscopy experiments. SlowFade Gold Antifade Mountant is glycerol-based, with no mixing or curing required, which makes it ideal for immediate viewing of the sample. It comes ready-to-use—just apply a drop to the sample, add a coverslip, and image. It is available with or without DAPI nuclear stain.

Key attributes:
• Protects dyes from fading during imaging
• Ready-to-use liquid, ideal for immediate sample viewing
• Mounted samples are stable for weeks
• Maintains signal strength—little to no quenching
• Ideal for Alexa Fluor dyes

Select the antifade mounting or optical clearing reagent that matches your fluorescence imaging needs ›

SlowFade Gold Antifade Mountant is not recommended for mounting samples containing fluorescent proteins, such as GFP. For superior antifade protection of fluorescent proteins and fluorescent dyes in cultured cells or thin tissue slices, SlowFade Diamond Antifade Mountant is recommended. For high resolution or to image thicker tissue or 3D cell culture with a focal depth of 0–500 μm, try SlowFade Glass Antifade Mountant.

Select best SlowFade antifade reagent for your experimental needs ›

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineSlowFade
Quantity2 mL
Shipping ConditionRoom Temperature
For Use With (Equipment)Microscope
FormLiquid
Product TypeMounting Media
Solution TypeAnti-fade Mountant
Unit SizeEach
Contents & Storage
Storage at room temperature is recommended but can also be stored frozen (-5° to -30°C). Protect from light.

Frequently asked questions (FAQs)

Does Slowfade Antifade Mountant require using a nail polish for sealing?

No. Samples mounted with SlowFade mountants need not be sealed; they are intended for immediate viewing. The coverslip may be anchored (to prevent movement while viewing) by applying molten paraffin to three or four spots around the edge of the coverslip.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.