Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts
Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts.
Have Questions?
Change view
Catalog Number
Product Type
S4S-GK
Gaskets
R10384
Buffer Drain Bottle
STK-10
Stopcocks
S4S-DB
Buffer Drain Bottle
4 Options
Catalog number S4S-GK
Price (ZAR)
-
Product Type:
Gaskets
Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts, for use with the Owl™ S4S Aluminum-Backed Sequencer System.
Choose from these replacement parts for optimal manual sequencing.
Specifications
DescriptionReplacement Gasket
For Use With (Equipment)Owl™ S4S Aluminum-Backed Sequencer System