SYTO™ 13 Green Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO, 250 μL - FAQs
SYTO™ 13 Green Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO, 250 μL - FAQs
View additional product information for SYTO™ Green Fluorescent Nucleic Acid Stains - FAQs (S32703, S7575, S7574, S7573, S7572, S7578, S7556, S34854, S7576, S34855, S7559)
This product does not have any FAQs available.