Streptavidin, Allophycocyanin, crosslinked, conjugate - FAQs
Streptavidin, Allophycocyanin, crosslinked, conjugate - FAQs
View additional product information for Streptavidin, Allophycocyanin, crosslinked, conjugate - FAQs (S868)
1 product FAQs found
How should I store my allophycocyanin (APC)-conjugated antibody?
Allophycocyanin-conjugated antibodies must be stored at 2-8 degrees C. Antibodies conjugated to phycobiliproteins (APC, R-PE) must never be frozen. Once frozen, even briefly, they are no longer usable.