Streptavidin Agarose, sedimented bead suspension - FAQs
Streptavidin Agarose, sedimented bead suspension - FAQs
View additional product information for Streptavidin Agarose, sedimented bead suspension - FAQs (S951)
1 product FAQs found
What is the binding capacity of streptavidin agarose?
The binding capacity of streptavidin agarose is 15-20 µg (18-24 nmoles) of fluorescein biotin per mL of sedimented gel.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.