Streptavidin, (FITC)
Invitrogen™

Streptavidin, (FITC)

• Class: RUO• Statement: For research use only. Not for use in diagnostic procedures.• Buffer: PBS• Preservative: 0.1% Sodium AzideViewRead more
Have Questions?
Catalog NumberQuantity
SA10011 mL
Catalog number SA1001
Price (USD)
248.00
Each
Quantity:
1 mL
Price (USD)
248.00
Each
• Class: RUO
• Statement: For research use only. Not for use in diagnostic procedures.
• Buffer: PBS
• Preservative: 0.1% Sodium Azide

View a selection guide for all Alexa Fluor™ streptavidin and avidin conjugates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferPBS Buffer
Detection MethodFluorescence
Excitation/Emission488/525
Flow Cytometer Laser Lines488
For Use With (Equipment)Flow Cytometer
Label or DyeFITC (Fluorescein)
PackagingTube
Product TypeStreptavidin Conjugate (Fluorescent)
Quantity1 mL
Shipping ConditionWet Ice
ConjugateFITC (Fluorescein)
FormSolution
Unit SizeEach
Contents & Storage
Contains 1 vial of streptavidin, FITC conjugate.

Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

Where can I find the antibody concentration for Streptavidin, (FITC) (Cat. No. SA1001)?

The antibody concentration for Streptavidin, (FITC) can be found in the Certificate of Analysis for your specific lot number.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.