Streptavidin, (Cy5)
Invitrogen™

Streptavidin, (Cy5)

• Class: RUO• Statement: For research use only. Not for use in diagnostic procedures.• Buffer: PBS• Preservative: 0.1% Sodium AzideViewRead more
Have Questions?
Catalog NumberQuantity
SA10110.5 mL
Catalog number SA1011
Price (USD)
192.00
Each
Quantity:
0.5 mL
Price (USD)
192.00
Each
• Class: RUO
• Statement: For research use only. Not for use in diagnostic procedures.
• Buffer: PBS
• Preservative: 0.1% Sodium Azide

View a selection guide for all Alexa Fluor™ streptavidin and avidin conjugates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferPBS Buffer
Detection MethodFluorescence
Excitation/Emission600-650/670
Flow Cytometer Laser Lines633/635
For Use With (Equipment)Flow Cytometer
Label or DyeCy5
PackagingTube
Product TypeStreptavidin Conjugate (Fluorescent)
Quantity0.5 mL
Shipping ConditionWet Ice
ConjugateCy5
FormSolution
Product LineCy
Unit SizeEach
Contents & Storage
Contains 1 vial of streptavidin, Cy5 conjugate.

Store in refrigerator (2–8°C).