Botte Assemblies now configurable in the Single Use Assembly Configurator Tool!
Standard Single-Use Bottle Assembly System, polycarbonate, 2-port cap set
Thermo Scientific™
Standard Single-Use Bottle Assembly System, polycarbonate, 2-port cap set
This 1-L Thermo Scientific polycarbonate (PC) Single-Use Bottle Assembly System includes a Nalgene Square Polycarbonate Bottle. It is ready toRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SB00003I
also known as SB00003-I
500 mL
SB00001I
also known as SB00001-I
125 mL
SB00002I
also known as SB00002-I
250 mL
SB00004I
also known as SB00004-I
1 L
Catalog number SB00003I
also known as SB00003-I
Price (USD)
-
Capacity (Metric):
500 mL
This 1-L Thermo Scientific polycarbonate (PC) Single-Use Bottle Assembly System includes a Nalgene Square Polycarbonate Bottle. It is ready to use and also includes a 38–430 mm ported closure with one vent filter line and one 1/8-in plugged connection line.
Specifications
Capacity (English)16.9 oz.
Capacity (Metric)500 mL
Closure MaterialHDPE
Closure TypeScrew Cap
MaterialPolycarbonate
Quantity500 mL, 1 case of 12
SterilitySterile
Through the Closure DispensingYes
Workflow StepUpstream sampling. Upstream Validation. Upstream process solution storage.
Product TypeSingle Use Mixer Transfer Assembly
Unit SizeCase of 12
Contents & Storage
Store at room temperature.