Lambda DNA - FAQs
Lambda DNA - FAQs
View additional product information for Lambda DNA - FAQs (SD0011)
2 product FAQs found
Which E. coli strain is lambda DNA (Cat. No. SD0011) isolated from?
Lambda DNA (Cat. No. SD0011) is isolated from a heat-inducible lysogenic E. coli W3110 strain.
What concentration is lambda DNA (Cat. No. SD0011) provided at?
Lambda DNA (Cat. No. SD0011) is provided at a concentration of 0.3 µg/µL.