CpG Methylated Human Genomic DNA
Thermo Scientific™

CpG Methylated Human Genomic DNA

Thermo Scientific Human genomic DNA is enzymatically methylated with CpG Methyltransferase (M.SssI). Genomic DNA was isolated from blood samples fromRead more
Have Questions?
Catalog NumberQuantity
SD113115 μg
Catalog number SD1131
Price (USD)
415.00
15 µg
Quantity:
15 μg
Price (USD)
415.00
15 µg
Thermo Scientific Human genomic DNA is enzymatically methylated with CpG Methyltransferase (M.SssI). Genomic DNA was isolated from blood samples from healthy male (HIV, HBV, and HCV negative) individuals. Isolated genomic DNA was treated with CpG Methyltransferase, repurified, and dissolved in TE buffer. ≥ 98% CpG methylation level confirmed by bisulfite sequencing.

Highlights

• Pure—DNA free from inhibitors, proteins and RNA
• Convenient—ready to use 100 ng/μL concentration
• Compatible with downstream applications
• Complete methylation of cytosines in CpG dinucleotides

Applications

As controls in the following:
• Bisulfite sequencing
• Combined Bisulfite Restriction Analysis (COBRA)
• Methylation-sensitive Single-Nucleotide Primer Extension (Ms-SNuPE)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration100 ng/μL
For Use With (Application)Sequencing
FormLiquid
Includes[other]
Product TypeCpG Methylated Human Genomic DNA
Quantity15 μg
SpeciesHuman
FormatTube
Unit Size15 µg