Please note: order placement quantity must be in multiples of 25, minimum of 25 required.
2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 2 ports, MPC insert connectors
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 2 ports, MPC insert connectors

Large two-port Thermo Scientific™ Labtainer™ BioProcess Containers (BPCs) are available in 5 L, 10 L, and 20 L for mixedRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SH00024I
also known as SH00024-I
20 L
SH00022I
also known as SH00022-I
5 L
SH00023I
also known as SH00023-I
10 L
Catalog number SH00024I
also known as SH00024-I
Price (USD)
-
Capacity (Metric):
20 L
Large two-port Thermo Scientific™ Labtainer™ BioProcess Containers (BPCs) are available in 5 L, 10 L, and 20 L for mixed flow applications. Port 1 has ¼-inch tubing for small flow applications while port 2 supports a higher flow with 3/8-inch tubing. These bags feature male MPC connectors for ease of use and security as well as a built-in bag hanger for better handling. The 10 L Thermo Scientific™ BPC 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL ethyl vinyl acetate film and includes 2 ports (2 MPC insert + barb and cap).

Features of the system include:
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications include:
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquid
• Collecting chromatography feed and fraction
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors
• Filters (used for bioburden reduction)—sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers and platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Specifications
Dimensions63.3 cm x 42.5 cm (24.92 in. X 16.75 in.)
Outer PackagingTwo 4-mil. poly bags
Product LineLabtainer
Quantity20 L, 25 per case
RegulatorycGMP
Shipping ConditionRoom Temperature
Sterilization MethodGamma Irradiated
Capacity (Metric)20 L
Film DescriptionASI 28
MaterialEVA
Port DetailMPC Insert Connector
Tubing StyleEVA
Unit Size20 L
Contents & Storage
Store at room temperature.