2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 1 port, Luer Lock body connector
Please note: order placement quantity must be in multiples of 50, minimum of 50 required.
2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 1 port, Luer Lock body connector
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 1 port, Luer Lock body connector

The 100 mL Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SH00038I
also known as SH00038-I
1 L
SH00035I
also known as SH00035-I
100 mL
SH00036I
also known as SH00036-I
250 mL
SH00037I
also known as SH00037-I
500 mL
SH00039I
also known as SH00039-I
2 L
Catalog number SH00038I
also known as SH00038-I
Price (USD)
-
Request A Quote
Capacity (Metric):
1 L
The 100 mL Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 1 port (1 luer body + cap).

Features of the system include:
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications include:
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquid
• Collecting chromatography feed and fraction
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors
• Filters (used for bioburden reduction)—sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers and platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Specifications
Dimensions28.6 cm x 13.3 cm (11.25 in. X 5.25 in.)
Outer PackagingTwo 4-mil. poly bags
Product LineLabtainer
Quantity1 L, 50 per case
RegulatorycGMP
Shipping ConditionRoom Temperature
Sterilization MethodGamma Irradiated
Capacity (Metric)1 L
Film DescriptionASI 28
MaterialEVA
Port DetailLuer Lock Body Connector
Tubing StyleEVA
Unit Size1 L
Contents & Storage
Store at room temperature.