HyPerforma™ 50 to 500L Tank Liners, 100L Liner - Bottom drain, irradiated
Thermo Scientific™
HyPerforma™ 50 to 500L Tank Liners, 100L Liner - Bottom drain, irradiated
The 100 L Thermo Scientific™ tank liner with 1 bottom drain is composed of CX3-9 single web, multi-layer plastic film,Read more
Have Questions?
Catalog NumberQuantity
SH3064602
also known as SH30646.02
Catalog number SH3064602
also known as SH30646.02
Price (MXN)
-
The 100 L Thermo Scientific™ tank liner with 1 bottom drain is composed of CX3-9 single web, multi-layer plastic film, with dimensions optimized for Thermo Scientific drums and commonly used industry-standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery.
Features:
• Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
• Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications of the Thermo Scientific tank liner:
• Removes the need for tank cleaning and helps reduce cycle times.
• Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
Specifications
For Use With (Equipment)HyPerforma Conical Bottom PE Drums - SV50517.05
Outer PackagingPlaced in strong cardboard cartons with product and lot identification.
Product LineHyPerforma
Product TypeTank Liner
Protective Inner PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
SterilitySterile
Sterilization MethodIrradiated
Capacity (Metric)100 L
Film DescriptionCX3-9
MaterialPlastic
Port Detail1/2 in. Quick Disconnect Insert (MPX series);MPX insert, polycarbonate