Labtainer™ BioProcess Container (BPC), 1L with 2 Ports, Luer Lock, 2D BPC
Thermo Scientific™

Labtainer™ BioProcess Container (BPC), 1L with 2 Ports, Luer Lock, 2D BPC

The 1 L Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single-web, multi-layer film, and includes 2 edgeRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Tubing Style
SH30657.151 LNo Tubing
SH3065815
also known as SH30658.15
1 L12 in. of 1/4 in. Tubing
Catalog number SH30657.15
Price (USD)
-
Capacity (Metric):
1 L
Tubing Style:
No Tubing
The 1 L Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single-web, multi-layer film, and includes 2 edge ports with luer lock body and insert connections.

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All Labtainers are constructed in an ISO 7 cleanroom under cGMP conditions.

Applications of the Thermo Scientific Labtainer BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors.
• Buffer, culture media, and process liquid preparation and storage.
• Chromatography feed and fraction collection.
• Harvest, storage and transport of bulk drug product and bulk drug precursors.

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library – with over 20,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
Filters—used for bioburden reduction; sterilization grade filters and vent filters.
Tubing—thermoplastic elastomers, platinum-cured silicone.
Sample ports—septum-style and needle-free ports.
Clamps—sliders, pinch, and crimp bands.

Specifications
Dimensions29.7 cm x 20.1 cm (11.7 in. x 7.9 in.)
Outer PackagingPackages placed in strong cardboard cartons with product and lot identification
Product LineLabtainer
Protective Inner PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity10 per case
Sterilization MethodGamma Irradiated
Tubing Line 1Line 1. Body luer lock connection, polypropylene. No Tubing
Tubing Line 2Line 2. Insert luer lock connection, polypropylene. No Tubing Line
Capacity (Metric)1 L
Film DescriptionCX5-14
Port DetailEdge Ports: 2 x 0.25 in.
Tubing StyleNo Tubing
Unit Size1 L