BioProcess Container (BPC), 200L, for Single-Use Mixing Systems (S.U.M.s), for use with liquid-to-liquid mixing applications
Thermo Scientific™
BioProcess Container (BPC), 200L, for Single-Use Mixing Systems (S.U.M.s), for use with liquid-to-liquid mixing applications
The 200L, liquid-to-liquid, Thermo Scientific™ BioProcess Container (BPC) for Single-Use Mixing Systems (S.U.M.) is a flexible container system with specially designed ports and connectors for compatibility with S.U.M.™ systems.
Have Questions?
Catalog NumberQuantity
SH3073401
also known as SH30734.01
200 L, each
Catalog number SH3073401
also known as SH30734.01
Price (USD)
-
Quantity:
200 L, each
The 200L, liquid-to-liquid, Thermo Scientific™ BioProcess Container (BPC) for Single-Use Mixing Systems (S.U.M.) is a flexible container system with specially designed ports and connectors for compatibility with S.U.M.™ systems.
BPC Components include:
  • Powder Addition Port for addition of powder intended for connection to Powdertainer
  • Ports for addition of media and other liquids
  • Seal/Bearing Assembly that is linked to seal/bearing assembly by C-Flex™ tubing which forms the contact material for the shaft
  • Impeller of molded polyethylene that is linked to seal/bearing assembly by C-Flex tubing which forms the contact material for the shaft
  • Drain Port used when draining the S.U.M.
Specifications
For Use With (Application)Liquid
Quantity200 L, each
Capacity (Metric)200 L
Unit Size200 L