PowderTainer™ II 1 to 25 kg Single-Use BPC for powder applications, 5 kg
Thermo Scientific™

PowderTainer™ II 1 to 25 kg Single-Use BPC for powder applications, 5 kg

The 5 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 film that includes 2 ports (1 wash down line, 1 powder port).
Have Questions?
Catalog NumberCapacity (Metric)
SH3073702
also known as SH30737.02
5 kg
Catalog number SH3073702
also known as SH30737.02
Price (USD)
-
Capacity (Metric):
5 kg

The 5 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 film that includes 2 ports (1 wash down line, 1 powder port). The Powdertainer BPC is specifically designed for powder containment and discharge applications, and maintains a closed system for maximum recovery of powder while minimizing the risk of cross contamination.

Features:

  • Three-inch tri-clamp port designed for secure connection to, and easy integration, with existing formulation vessels.
  • Powdertainer BPC features suspensor handle for support during discharge, as well as neck clamp to retain powder prior to discharge.
  • Use this system to eliminate post-use cleaning steps required with re-useable containers.

Applications of the Thermo Scientific™ Powdertainer BPC include:

  • Storage and delivery of powder culture media and buffers.
  • Storage and dispensing of pre-weighed chemicals and other process powders.
Specifications
Capacity (Metric)5 kg
Dimensions (L x W)(22.1 x 14.3 in.) 56.1 x 36.3 cm
Film DescriptionCX3-9
Includes2 Ports (1 Wash Down Line, 1 Powder Port)
Length (Metric)56.1 cm
Port DetailPowder port: 7.6 cm (3 in.) tri-clamp with integrated seal
Product LinePowderTainer™ II
Width (Metric)36.3 cm
Product TypeBioprocess Container for Powder
Width (English)14.3 in.