Product Image
Thermo Scientific™
BioProcess Container (BPC), 1000L, for HyPerforma™ Single-Use Mixer, open-top mixing liner
Have Questions?
Catalog NumberCapacity (Metric)
SH30762.03
also known as SH3076203
1000 L
Catalog number SH30762.03
also known as SH3076203
Price (USD)
-
Capacity (Metric):
1000 L
The 1000L standard, open-top tank liner, constructed of CX3-9 film, is for use with the Thermo Scientific™ HyPerforma™ Single-Use Mixer and docking tank for mixing applications.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Specifications
Film DescriptionCX3-9
Port Detail0 Probe Port
Product LineHyPerforma
Capacity (Metric)1000 L