PowderTainer™ II 1 to 25kg Single-Use BPC for powder applications, 25 kg
Thermo Scientific™

PowderTainer™ II 1 to 25kg Single-Use BPC for powder applications, 25 kg

The 25 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 filmRead more
Have Questions?
Catalog NumberPort Detail
SH3086403
also known as SH30864.03
Powder Port: 3 in.
Catalog number SH3086403
also known as SH30864.03
Price (USD)
-
Port Detail:
Powder Port: 3 in.
The 25 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 film that includes 1 port (1 powder port). The Powdertainer BPC is specifically designed for powder containment and discharge applications, and maintains a closed system for maximum recovery of powder while minimizing the risk of cross contamination.

Features:
• Three-inch tri-clamp port designed for secure connection to, and easy integration, with existing formulation vessels.
• Powdertainer BPC features suspensor handle for support during discharge, as well as neck clamp to retain powder prior to discharge.
• Use this system to eliminate post-use cleaning steps required with re-useable containers.

Applications of the Thermo Scientific Powdertainer BPC include:
• Storage and delivery of powder culture media and buffers.
• Storage and dispensing of pre-weighed chemicals and other process powders.

Specifications
Dimensions (L x W x H)83.3 x 59.2 cm (32.8 x 23.3 in.)
Film DescriptionCX3-9
Includes1 Port (1 Powder Port)
Length (Metric)83.3 cm
Outer PackagingPackages are placed in strong cardboard cartons with product and lot identification.
Port DetailPowder Port: 3 in.
Product LinePowderTainer™ II
Protective Inner PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area.
Sterilization MethodNon-Irradiated
Tubing Line 1Washdown Line: N/A
Tubing Line 2Powder port: 7.6 cm (3 in.) tri-clamp with intergrated seal
Weight (Metric)25 kg
Width (Metric)59.2 cm
Product TypeBioprocess Container for Powder
Width (English)23.3 in.