Dolly for 300L Plastic Drum for HyPerforma™ Single-Use Mixer DS 300 (stainless steel)
Thermo Scientific™
Dolly for 300L Plastic Drum for HyPerforma™ Single-Use Mixer DS 300 (stainless steel)
The Dolly (single pack) is designed for use with 300L Plastic Drums and the Thermo Scientific™ HyPerforma™ Single-Use Mixer (S.U.M.) DS 300. Dollies for the drums are available in two sizes: one for 50 to 200L drums and another for 300L drums. These drums should be used with the corresponding optional drum positioner that is available with the docking station.
Have Questions?
Catalog NumberQuantity
SH30958.031 Ea.
Catalog number SH30958.03
Price (USD)
-
Quantity:
1 Ea.
The Dolly (single pack) is designed for use with 300L Plastic Drums and the Thermo Scientific™ HyPerforma™ Single-Use Mixer (S.U.M.) DS 300. Dollies for the drums are available in two sizes: one for 50 to 200L drums and another for 300L drums. These drums should be used with the corresponding optional drum positioner that is available with the docking station.
Required accessories:
  • HyPerforma Single-Use Mixer DS 300 docking station
  • HyPerforma Single-Use Mixer DS 300 bioprocess container bags for closed-top mixing or liners for open-top mixing
Recommended for:
  • Powder-liquid or liquid-liquid mixing applications in single-use
  • High-throughput mixing and storage
Specifications
Capacity (Metric)300 L
MaterialPolyethylene
Product LineHyPerforma
Quantity1 Ea.
For Use With (Equipment)DS 300 Single-Use Mixer
Unit Size1 pack