Thermo Scientific™ Single-Use Fermentor BioProcess Container, CX5-14 film, pH/DO sensor, foam sensor, Meissner inlet, four 10-inch exhaust filters, 300 L
Thermo Scientific™
Thermo Scientific™ Single-Use Fermentor BioProcess Container, CX5-14 film, pH/DO sensor, foam sensor, Meissner inlet, four 10-inch exhaust filters, 300 L
This 300 L Thermo Scientific™ Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of CX5-14 film, a five-layer, animal-origin free film, and includes a Mettler Toledo single-use pH and DO sensor, foam sensor, Meissner inlet, four 10-inch exhaust filters, and condenser.
Have Questions?
Catalog NumberQuantity
SH3101703300 L
Catalog number SH3101703
Price (USD)
-
Quantity:
300 L
This 300 L Thermo Scientific™ Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of CX5-14 film, a five-layer, animal-origin free film, and includes a Mettler Toledo single-use pH and DO sensor, foam sensor, Meissner inlet, four 10-inch exhaust filters, and condenser.
All SUF BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
Features:
  • Manufactured with industry-leading CX5-14 film
  • C-flex tubing that can be used with tubing welder and various connection options
  • Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from the industry's largest component library, with over 2,000 unique components that include:
  • Fittings – straight, reducing, elbow, T-style, X-style, and cross-style
  • Connectors – quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
  • Filters – used for bioburden reduction; sterilization grade filters and gas filters
  • Tubing – thermoplastic elastomers, platinum-cured silicone
  • Sample ports – septum-style and needle-free ports
  • Clamps – sliders, pinch, and crimp bands
Specifications
Product LineHyPerforma
Quantity300 L
Capacity (Metric)300 L
Unit Size300 L
Contents & Storage
Store at room temperature.