Labtainer™ 2D BioProcess Container (BPC), Aegis™ 5-14 film, three ports, Luer locks
Thermo Scientific™
Labtainer™ 2D BioProcess Container (BPC), Aegis™ 5-14 film, three ports, Luer locks
Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis 5-14 single-web, multi-layer film.Features of the Labtainer 2D BPCWeitere Informationen
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerKapazität (metrisch)
SH310490450 l
SH310501150 ml
SH3105012100 ml
SH3105013250 ml
SH3105014500 ml
SH31050151 l
SH31050162.000 ml
SH31049015 l
SH310490210 l
SH310490320 l
SH31049052 l
Katalognummer SH3104904
Preis (EUR)
-
Kapazität (metrisch):
50 l
Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis 5-14 single-web, multi-layer film.
Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions
Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
Specifications
Kapazität (metrisch)50 l
Zertifizierung/KonformitätISO 7
FilmbeschreibungAegis 5-14
Zur Verwendung mit (Anwendung)Bioprocessing
Anschlussdetail3 ports
Menge50 L
SchlauchstilC-Flex
AutoklavierbarNicht autoklavierbar
FarbeDurchsichtig
EinwegproduktSingle-use
SterilitätSteril
Unit Size50 L