Harvestainer™ Microcarrier Separation System
Thermo Scientific™
Harvestainer™ Microcarrier Separation System
The Thermo Scientific™ Harvestainer Microcarrier Separation System is the ideal solution for the separation of microcarrier beads during adherent cell culture production.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SH3107802
also known as SH31078.02
12 L
SH31078013 L
SH3107101
also known as SH31071.01
25 L
SH3107102
also known as SH31071.02
50 L
Catalog number SH3107802
also known as SH31078.02
Price (USD)
-
Capacity (Metric):
12 L
The Thermo Scientific™ Harvestainer Microcarrier Separation System is the ideal solution for the separation of microcarrier beads during adherent cell culture production. The Harvestainer system is a closed, single-use system that helps to increase product yields versus traditional methods, while reducing clean-in-place and steam-in-place requirements and water for injection usage. It eliminates equipment maintenance and reduces process validations, cycle times, and manual final decanting processes.
For smaller-scale or larger-scale applications, the Harvestainer system is ideal for in-line microcarrier separation. The 3 L and 12 L Harvestainer systems consist of a pre-assembled bioprocess container (BPC) and tray in a single-use unit, where the tray acts as the secondary containment device. The 25 L and 50 L Harvestainer systems consist of a 3D BPC with a drum as the secondary containment device. The BPC features a dip tube with dipwell design for better drainage and minimal manipulation.
  • Closed system
  • Scalable for both smaller-scale and larger-scale applications
  • Helps deliver high recovery rates
  • Inline microcarrier separation
Applications: Adherent cell culture production
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity (Metric)12 L
MaterialPETG
Product LineHarvestainer
Quantity12 L
Shipping ConditionRoom Temperature
SterilitySterile
Unit Size12 L
Contents & Storage
Room Temperature