imPULSE™ Single-Use Mixer and BioProcess Containers, ASI™ 26/77 Film
Thermo Scientific™
imPULSE™ Single-Use Mixer and BioProcess Containers, ASI™ 26/77 Film
The Thermo Scientific imPULSE Single-Use Mixer BioProcess Container (BPC) aids in providing greater process efficiency for most upstream and downstream process operations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Port Detail
SH313683000 LWith Ports
SH31358100 LWith Ports
SH31357100 LWithout Ports
SH31360250 LWith Ports
SH31359250 LWithout Ports
SH31362500 LWith Ports
SH31361500 LWithout Ports
SH313641000 LWith Ports
SH313631000 LWithout Ports
SH313662000 LWith Ports
SH313652000 LWithout Ports
SH313673000 LWithout Ports
SH313705000 LWith Ports
SH313695000 LWithout Ports
Catalog number SH31368
Price (KRW)
-
Capacity (Metric):
3000 L
Port Detail:
With Ports
The Thermo Scientific imPULSE Single-Use Mixer BioProcess Container (BPC) is designed for use with the imPULSE Single-Use Mixer, and is used for both upstream and downstream mixing applications. Together, these products help deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
  • Designed for use with the imPULSE Single-Use Mixer
  • Used for both upstream and downstream mixing applications
  • Together, these products help deliver simplified connectivity and enable greater process efficiency
Specifications
Description3000 L imPULSE Single-Use Mixer (S.U.M.) BioProcess Container (BPC), ASI 26/77 film with ports
Film DescriptionASI 26/77 Film
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
Port DetailWith Ports
QuantityEach
Capacity (Metric)3000 L
Unit Size3000 L
Contents & Storage
Ambient/room temperature