phiX174 DNA/BsuRI (HaeIII) Marker, 50 μg - FAQs
phiX174 DNA/BsuRI (HaeIII) Marker, 50 μg - FAQs
View additional product information for phiX174 DNA/BsuRI (HaeIII) Marker - FAQs (SM0252, SM0251)
1 product FAQs found
What is the recommended voltage for electrophoresis with Thermo Scientific DNA and RNA ladders? What does V/cm mean?
Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).