GeneRuler DNA Ladder Mix, ready-to-use, 100 applications, 100 to 1000 bp, 21 fragments - FAQs
GeneRuler DNA Ladder Mix, ready-to-use, 100 applications, 100 to 1000 bp, 21 fragments - FAQs
View additional product information for GeneRuler Ready-to-Use DNA Ladders - FAQs (SM1193, SM0333, SM0244, SM1333, SM0324, SM0313, SM1213, SM1334, SM0323, SM0334, SM0373, SM1353, SM0243, SM0314)
4 product FAQs found
Are the Thermo Scientific DNA ladders compatible with GelRed and GelGreen gel stains?
The GelRed or GelGreen nucleic acid dyes can be used with our DNA Ladders. Nucleic acid binding dyes can affect DNA migration during electrophoresis, therefore we highly recommend post-staining of gels. Please follow the gel staining protocol provided by the manufacturer of nucleic acid gel dyes.
Which Thermo Scientific DNA ladders are optimal for fast electrophoresis separation?
For fast sizing and quantification of double-stranded DNA, we recommend using FastRuler DNA Ladders. These ladders are easily resolved in a short separation distance after an 8-14 min run.
What is the recommended voltage for electrophoresis with Thermo Scientific DNA and RNA ladders? What does V/cm mean?
Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).
What are the end functional groups of Thermo Scientific DNA and RNA ladders?
Thermo Scientific DNA and RNA ladders have 5′-phosphate and 3′-hydroxyl groups.