Savant™ SpeedVac™ Integrated Vacuum Concentrator Systems and Kits
Savant™ SpeedVac™ Integrated Vacuum Concentrator Systems and Kits
Savant™ SpeedVac™ Integrated Vacuum Concentrator Systems and Kits
Savant™ SpeedVac™ Integrated Vacuum Concentrator Systems and Kits
Thermo Scientific™

Savant™ SpeedVac™ Integrated Vacuum Concentrator Systems and Kits

Get all the necessary components for HPLC fractions with water/0.1% trifluoroacetic acid and reversed-phase SPE eluants applications with these kits.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelIncludesElectrical Requirements
SPD1030-115SPD1030Vacuum Concentrator115 V 60 Hz
SPD1030-230SPD1030Vacuum Concentrator230 V 50 Hz
SPD1030A-115SPD1030Diaphragm Pump, Cold Trap, Vacuum Gauge115 V 60 Hz
SPD1030A-230SPD1030Diaphragm Pump, Cold Trap, Vacuum Gauge, RH40-11 Rotor230 V 50 Hz
SPD1030P1-115SPD1030Diaphragm Pump, Cold Trap, Vacuum Gauge, RH40-11 Rotor115 V 60 Hz
SPD1030P1-230SPD1030Diaphragm Pump, -50°C Cold Trap, Vacuum Gauge, RH40-11 Rotor230 V 50 Hz
SPD2030-220SPD2030Diaphragm Pump, Cold Trap, Vacuum Gauge220 V 60 Hz
SPD2030-230SPD2030Diaphragm Pump, Cold Trap, Vacuum Gauge230 V 50 Hz
SPD2030A-220SPD2030Diaphragm Pump, Cold Trap, Vacuum Gauge220 V 60 Hz
SPD2030A-230SPD2030Diaphragm Pump, Cold Trap, Vacuum Gauge230 V 50 Hz
SPD2030P1-220SPD2030Diaphragm Pump, -50°C Cold Trap, Vacuum Gauge, RH40-11 Rotor220 V 60 Hz
SPD2030P1-230SPD2030Diaphragm Pump, -50°C Cold Trap, Vacuum Gauge, RH40-11 Rotor230 V 50 Hz
Catalog number SPD1030-115
Price (USD)
-
Model:
SPD1030
Includes:
Vacuum Concentrator
Electrical Requirements:
115 V 60 Hz

Dry and concentrate aqueous or non-aggressive samples with speed and efficiency with Thermo Scientific™ Savant SPD1030 and SPD2030 Integrated SpeedVac™ Systems and kits. Fully integrated for fast installation, programming. Ideal for drying DNA/RNA precipitates, PCR samples, proteins, enzyme, HPLC fractions in plates or tubes in water/ethanol or isopropanol and other organic solvents. SpeedVac SPD1030 System is designed for medium capacity (i.e.,120 x 1.5 mL tubes, other formats). SpeedVac SPD2030 System allows higher capacity (i.e.,200 x 1.5 mL tubes, other formats).

Easy to Use:

  • Plug and play installation: Integrated solution includes a concentrator, oil-free vacuum pump and cold trap all integrated in one unit. No tubing connections needed.
  • Intuitive and user friendly program digital display which allows use of three preset programs and ability to create three customizable programs to set heat, run time, ramp rate and vacuum level for efficient operation.
  • Wide variety of rotors to support sample format from micro tubes, vials and microplates to 250-ml flasks
  • Audible alarm to alert user when operation is finished
  • Kits are available, which includes the integrated system and rotor

Robust, Low-Maintenance Design:

  • Oil-free vacuum pump is virtually maintenance free
  • PTFE coated chamber protects the chamber from corrosion, while making cleaning easy

Enhanced Evaporation:

  • Faster sample drying provided with radiant cover
  • Vacuum ramping and pressure controls eliminate sample bumping to maintain sample integrity
  • Reduced vapor path from concentrator to refrigerated vapor trap accelerates evaporation
  • Rotors fit a wide variety of sample formats such as tubes, flasks, and shallow-well and deep-well microplates

Solvent and Solvent Combinations:

  • Acetonitrile
  • Ethanol
  • Methanol
  • Methylene Chloride
  • Water
  • Other organic solvents

 

SPD1030115 and SPD1030230 includes:
  • Concentrators only, accessories sold separately

SPD1030A and SPD2030A includes:

  • Integrated SpeedVac system
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)

SPD1030P1 Kit includes:

  • Fully Integrated SpeedVac system
  • 40 × 1.5mL rotor (RH40-11)
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)

SPD2030P1 kit

  • Integrated SpeedVac system
  • 200 × 1.5mL rotor (RH200-12)
  • CryoCool™ 1L heat transfer fluid
  • Glass condensation Flask (GCF400)
Specifications
Amperage8 A
CertificationsUL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
DescriptionSPD1030 Integrated Vacuum Concentrator
Dimensions (L x W x H)25 x 26 x 16 in. (64 x 66 x 41 cm)
Electrical Requirements115 V 60 Hz
HoldsAccommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
IncludesVacuum Concentrator
Max. Vacuum<10 Torr (13 mbar, 1.3k Pa)
Shipping Weight (English)146 lb.
Shipping Weight (Metric)86 kg
Weight (Metric)66 kg
Chamber MaterialPTFE-coated Aluminum
ModelSPD1030
Temperature Range (Metric)Ambient to 80°C
Unit SizeEach