Savant™ SpeedVac™ SPD300 : Large Capacity Vacuum Concentrator and Kits
Thermo Scientific™

Savant™ SpeedVac™ SPD300 : Large Capacity Vacuum Concentrator and Kits

Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.

製品番号(カタログ番号)概要電気的要件
SPD300P2-230SpeedVac SPD300DDA Vacuum Concentrator Kit230 V 50 Hz
SPD300DDAA-115SpeedVac SPD300DDA Vacuum Concentrator Kit-No rotor115 V 60 Hz
SPD300DDAA-230SpeedVac SPD300DDA Vacuum Concentrator Kit-No rotor230 V 50 Hz
SPD300P1-115SpeedVac SPD300DDA Vacuum Concentrator Kit115 V 60 Hz
SPD300P1-230SpeedVac SPD300DDA Vacuum Concentrator Kit230 V 50 Hz
製品番号(カタログ番号) SPD300P2-230
価格(JPY)
-
概要:
SpeedVac SPD300DDA Vacuum Concentrator Kit
電気的要件:
230 V 50 Hz

Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.

  • For strong acids and bases used in combinatorial chemistry
仕様
認証UL Listed to US and Canadian requirements. CE marked
概要SpeedVac SPD300DDA Vacuum Concentrator Kit
電気的要件230 V 50 Hz
Unit SizeEach