Oxoid™ Skirrow Campylobacter Selective Supplement
Thermo Scientific™
Oxoid™ Skirrow Campylobacter Selective Supplement
Oxoid Campylobacter Selective Supplement, Skirrow prepares Skirrow Selective Medium for isolation of Campylobacter spp. at 42°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberYield
SR0069EEach vial for 500mL medium
SR0069HEach vial for 2L medium
Catalog number SR0069E
Price (USD)
-
Yield:
Each vial for 500mL medium
Oxoid Campylobacter Selective Supplement, Skirrow is used for preparation of Skirrow Selective Medium for the isolation of Campylobacter spp. at 42°C.
Add to Blood Agar Base No. 2, Part No. CM0271B or Columbia Blood Agar Base, Part No. CM0331B
Specifications
DescriptionCampylobacter Selective Supplement, Skirrow
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
Unit SizeEach